Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Auramine O, 80% dye content

Catalog No. AAJ6034314
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6034314 25 g
AAJ6034318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6034314 Supplier Thermo Scientific Chemicals Supplier No. J6034314
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Fluorescent stain for acid-resistant bacteria in sputum | CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol

Fluorescent stain for acid-resistant bacteria in sputum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-27-2
Molecular Formula C17H22ClN3
Molecular Weight (g/mol) 303.834
MDL Number MFCD00012484
InChI Key KSCQDDRPFHTIRL-UHFFFAOYSA-N
Synonym C.I. 41000
PubChem CID 17170
ChEBI CHEBI:51876
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl

Specifications

Melting Point 250°C (decomposition)
UN Number UN2811
Beilstein 4030061
Solubility Information Soluble in water or ethanol
Formula Weight 303.84
Color Yellow
Quantity 25 g
Physical Form Powder
Chemical Name or Material Auramine O, 80% dye content
Hazard Category H302-H311-H351
Hazard Statement GHS H Statement
H311-H351-H302
Toxic in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 219-567-2
RTECSNumber BY3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.