Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Auramine O, Certified, C.I. DcAu-4 25 GR
SDP

Catalog No. 50980185
Encompass_Preferred

(Pyoctanunum aureum; Basic Yellow 2; Pyoktanin Yellow; Canary Yellow)
CAS #2465-27-2 C₁₇H₂₁N₃HCl·H₂O F.W. 321.86
Solubility: 1.0% Water; 4.0% Alcohol; 1.2% Cellosolve; 1.75% Glycol; 0.05% Xylene

Catalog No. 50-980-185 Supplier Electron Microscopy Sciences Supplier No. 10850
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(Pyoctanunum aureum; Basic Yellow 2; Pyoktanin Yellow; Canary Yellow)
CAS #2465-27-2 C₁₇H₂₁N₃HCl·H₂O F.W. 321.86
Solubility: 1.0% Water; 4.0% Alcohol; 1.2% Cellosolve; 1.75% Glycol; 0.05% Xylene

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.