Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Auramine O, MP Biomedicals™

Catalog No. ICN207741 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN207741 10 g
ICN19506450 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN207741 Supplier MP Biomedicals, Inc Supplier No. 0520774110

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2465-27-2
Molecular Formula C17H22ClN3
Molecular Weight (g/mol) 303.834
InChI Key KSCQDDRPFHTIRL-UHFFFAOYSA-N
Synonym C.I. 41000, 4, 4'-(imidocarbonyl)Bis(N, N'-dimethyl-amine) monohydrochloride
PubChem CID 17170
ChEBI CHEBI:51876
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl

Specifications

Formula Weight 303.8
Color Yellow
Quantity 10 g
Physical Form Powder
Chemical Name or Material Auramine O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.