Learn More
Selleck Chemical LLC Auranofin-5mg
SDP
Supplier:  Selleck Chemical LLC S43075MG
Encompass_Preferred
Auranofin is an inhibitor of thioredoxin reductase (TrxR) with IC50 of 88 nM for purified H. pylori TrxR in cell-free assay. Auranofin has anti-cancer activity and can completely inhibit bacterial growth at 1.2 μM. Auranofin is an FDA-approved gold-containing compound used for the treatment of rheumatoid arthritis.
Catalog No. 50-238-9681