Learn More
TARGETMOL CHEMICALS INC Aurantiamide acetate 1MG
SDP
Catalog No. 502880197
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Aurantiamide acetate (Asperglaucide) was isolated from the fermentation broth of Aspergillus penicilloides for the first time. Aurantiamide acetate is a selective and orally active cathepsin inhibitor. Aurantiamide acetate has anti-inflammatory activities and can be used for the study of inflammatory diseases[1][2]. Purity 99.68%
Catalog No. 50-288-0197 Supplier TARGETMOL CHEMICALS INC Supplier No. TJS22161MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Aurantiamide acetate (Asperglaucide) was isolated from the fermentation broth of Aspergillus penicilloides for the first time. Aurantiamide acetate is a selective and orally active cathepsin inhibitor. Aurantiamide acetate has anti-inflammatory activities and can be used for the study of inflammatory diseases[1][2]. Purity 99.68%