Learn More
Medchemexpress LLC Aurantiamide acetate | 56121-42-7 | MFCD17214814 | 99.9% | 444.52 g·mol⁻¹ | C27H28N2O4 | 10 MG
SDP
Catalog No. 5000289133
Encompass_Preferred
Aurantiamide acetate is a small-molecule cathepsin inhibitor used as a research reagent with reported anti-inflammatory activity. It inhibits cathepsin L (IC50 12 μM) and cathepsin B (IC50 49 μM), is supplied as a white to off-white solid, and is reported at ~99.9% purity (CAS 56121-42-7).
  • Selective cathepsin L and B inhibitor (IC50 12 μM and 49 μM).
  • Demonstrated anti-inflammatory activity for disease research.
  • High purity suitable for analytical and preclinical use.
  • Soluble in DMSO (50 mg/mL) and formulatable for in vivo dosing.
  • Stable when stored under recommended conditions for powder and solutions.
Catalog No. 50-002-89133 Supplier Medchemexpress LLC Supplier No. HYN290510MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Aurantiamide acetate is a small-molecule cathepsin inhibitor used as a research reagent with reported anti-inflammatory activity. It inhibits cathepsin L (IC50 12 μM) and cathepsin B (IC50 49 μM), is supplied as a white to off-white solid, and is reported at ~99.9% purity (CAS 56121-42-7).
  • Selective cathepsin L and B inhibitor (IC50 12 μM and 49 μM).
  • Demonstrated anti-inflammatory activity for disease research.
  • High purity suitable for analytical and preclinical use.
  • Soluble in DMSO (50 mg/mL) and formulatable for in vivo dosing.
  • Stable when stored under recommended conditions for powder and solutions.