Learn More
AdipoGen Aurantimycin A
SDP
Supplier:  AdipoGen BVT0398M001
Encompass_Preferred
Chemical. CAS 162478-50-4. Formula C38H64N8O14. MW 857. Isolated from Streptomyces aurantiacus JA 4570. Depsipeptide antibiotic. Active against Gram-positive bacteria. Cytotoxic agent. Nematocide activity. Antiulcer agent. Anti-inflammatory drug C5a antagonist.
Catalog No. 50-256-1949