Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aurantio-obtusin β-D-glucoside | 129025-96-3 | 1 MG
SDP

Catalog No. 5000420673
Encompass_Preferred

Aurantio-obtusin β-D-glucoside (Glucoaurantio-obtusin) is a glucoside of aurantio-obtusin, isolated from Cassiae Semen (seeds of Cassia tora). It appears as a yellow to brown solid with a purity of 99.43%. This product is intended for laboratory research and manufacture of substances, and is for research use only by experienced personnel.

  • Isolated from cassiae semen (seeds of cassia tora)
  • Glucoside of aurantio-obtusin
  • High purity of 99.43%
  • Appears as a yellow to brown solid
  • Intended for laboratory research and manufacture of substances
  • Stable under recommended storage conditions
  • Molecular weight: 492.43
  • Chemical formula: C23H24O12

Catalog No. 50-004-20673 Supplier Medchemexpress LLC Supplier No. HYN41791MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aurantio-obtusin β-D-glucoside (Glucoaurantio-obtusin) is a glucoside of aurantio-obtusin, isolated from Cassiae Semen (seeds of Cassia tora). It appears as a yellow to brown solid with a purity of 99.43%. This product is intended for laboratory research and manufacture of substances, and is for research use only by experienced personnel.

  • Isolated from cassiae semen (seeds of cassia tora)
  • Glucoside of aurantio-obtusin
  • High purity of 99.43%
  • Appears as a yellow to brown solid
  • Intended for laboratory research and manufacture of substances
  • Stable under recommended storage conditions
  • Molecular weight: 492.43
  • Chemical formula: C23H24O12

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.