Learn More
TARGETMOL CHEMICALS INC AURAPTENE 100MG
SDP
Catalog No. 502576808
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Auraptene is a coumarin derived from citrus plants that bears a geranyloxyl moiety at its C-7. It has anti-inflammatory anti-carcinogenic anti-bacterial neuroprotective and hepatoprotective activities. It inhibits leukocyte activation and induces phase II enzymes during the initiation phase of carcinogenesis.1 When examined for its potential use in Alzheimer's disease treatment auraptene was shown to inhibit (beta)-secretase (BACE1) activity with an IC50 value of 345.1 uM. purity: 99%
Catalog No. 50-257-6808 Supplier TARGETMOL CHEMICALS INC Supplier No. T4115100MG
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Auraptene is a coumarin derived from citrus plants that bears a geranyloxyl moiety at its C-7. It has anti-inflammatory anti-carcinogenic anti-bacterial neuroprotective and hepatoprotective activities. It inhibits leukocyte activation and induces phase II enzymes during the initiation phase of carcinogenesis.1 When examined for its potential use in Alzheimer's disease treatment auraptene was shown to inhibit (beta)-secretase (BACE1) activity with an IC50 value of 345.1 uM. purity: 99%