Learn More
Selleck Chemical LLC Auraptene S9052-1MG
SDP
Catalog No. 5000025351
Encompass_Preferred
Auraptene (7-geranyloxycoumarin) is a natural coumarin derived from citrus plants and possesses valuable pharmacological properties including anticarcinogenic anti-inflammatory antihelicobacter antigenotoxic and neuroprotective effects Auraptene inhibits matrix metalloproteinase 2 (MMP-2) as well as key inflammatory mediators including IL-6 IL-8 and chemokine (C-C motif) ligand-5(CCL5)
Catalog No. 50-000-25351 Supplier Selleck Chemical LLC Supplier No. S90521MG
Auraptene (7-geranyloxycoumarin) is a natural coumarin derived from citrus plants and possesses valuable pharmacological properties including anticarcinogenic anti-inflammatory antihelicobacter antigenotoxic and neuroprotective effects Auraptene inhibits matrix metalloproteinase 2 (MMP-2) as well as key inflammatory mediators including IL-6 IL-8 and chemokine (C-C motif) ligand-5(CCL5)