Learn More
Lkt Laboratories Auraptene, ≥98%, 25MG
SDP
Supplier:  Lkt Laboratories A807025MG
Encompass_Preferred
Monoterpene coumarin ether found in Citrus; PPARα agonist, ACAT inhibitor. CAS: 495-02-3. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4870