Learn More
Takara Bio Aureobasidin A, 10mg
SDP
Catalog No. NC0616072
Encompass
Aureobasidin A (AbA) is a cyclic depsipeptide antibiotic, isolated from the filamentous fungus Aureobasidium pullulans R106, which is toxic to yeast at low concentrations (0.10.5 g/ml). Sensitive species include Saccharomyces cerevisiae, Schizosaccharomyces pombe, Candida glabrata, Aspergillus nidulans and A. niger. AbA inhibits a yeast enzyme, inositol phosphorylceramide (IPC) synthase, which is expressed from the AUR1 gene. Expression of a mutant gene, AUR1-C, in transformed yeast confers resistance to the drug.
Catalog No. NC0616072 Supplier Takara Bio Supplier No. 630499
Aureobasidin A (AbA) is a cyclic depsipeptide antibiotic, isolated from the filamentous fungus Aureobasidium pullulans R106, which is toxic to yeast at low concentrations (0.10.5 g/ml). Sensitive species include Saccharomyces cerevisiae, Schizosaccharomyces pombe, Candida glabrata, Aspergillus nidulans and A. niger. AbA inhibits a yeast enzyme, inositol phosphorylceramide (IPC) synthase, which is expressed from the AUR1 gene. Expression of a mutant gene, AUR1-C, in transformed yeast confers resistance to the drug.