Learn More
TARGETMOL CHEMICALS INC Aureobasidin A 1MG
SDP
Catalog No. 502880205
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aureobasidin A is a cyclic peptide and an antifungal antibiotic. Aureobasidin A is an inhibitor of the inositolphosphorylceramide synthase AUR1. Purity 99.79%
Catalog No. 50-288-0205 Supplier TARGETMOL CHEMICALS INC Supplier No. T92671MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Aureobasidin A is a cyclic peptide and an antifungal antibiotic. Aureobasidin A is an inhibitor of the inositolphosphorylceramide synthase AUR1. Purity 99.79%