Learn More
Medchemexpress LLC HY-P1975 1mg , Aureobasidin A CAS:127785-64-2 Purity:>98%
SDP
Catalog No. 502259410
Encompass_Preferred
HY-P1975 1mg Aureobasidin A CAS: 127785-64-2 Aureobasidin A (Basifungin), a cyclic depsipetide, is an antifungal antibiotic. Aureobasidin A (Basifungin) A is an inhibitor of the inositolphosphorylceramide synthase AUR1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9410 Supplier Medchemexpress LLC Supplier No. HYP19751MG
HY-P1975 1mg Aureobasidin A CAS: 127785-64-2 Aureobasidin A (Basifungin), a cyclic depsipetide, is an antifungal antibiotic. Aureobasidin A (Basifungin) A is an inhibitor of the inositolphosphorylceramide synthase AUR1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.