Learn More
AdipoGen Aureothin
SDP
Supplier:  AdipoGen BVT0303M001
Encompass_Preferred
Chemical. CAS 2825-00-5. Formula C22H23NO6. MW 397.4. Isolated from Streptomyces thioluteus. Oxidoreductase inhibitor. Antitrypanosomal, antibacterial, antifungal, insecticidal and pesticidal. Antitumor compound.
Catalog No. 50-256-2006