Learn More
Abcam Aureothin, Tr x R1 inhibitor, 1MG
Catalog No. 502706000
Encompass_Preferred
MW 397.4 Da, Purity >96%. TrxR1 inhibitor. Broad spectrum antibiotic agent. Antiproliferative agent. Shows antitumor, antifungal, insecticidal, antibacterial and antiparasitic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6000 Supplier Abcam Supplier No. AB1441961MG
MW 397.4 Da, Purity >96%. TrxR1 inhibitor. Broad spectrum antibiotic agent. Antiproliferative agent. Shows antitumor, antifungal, insecticidal, antibacterial and antiparasitic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement