Learn More
AdipoGen Aureothricin
SDP
Catalog No. 502561977
Encompass_Preferred
Chemical. CAS 574-95-8. Formula C9H10N2O2S2. MW 242.3. Isolated from Streptomyces thioluteus. Antibiotic. Active against Gram-positive and Gram-negative bacteria, yeast, filamentous fungi, protozoa and insects. Potent bacterial and yeast RNA polymerases inhibitor. Inhibitor of fungal mannan and glucan formation. Similar to thiolutin. Antitumor compound.
Catalog No. 50-256-1977 Supplier AdipoGen Supplier No. BVT0345M001
Chemical. CAS 574-95-8. Formula C9H10N2O2S2. MW 242.3. Isolated from Streptomyces thioluteus. Antibiotic. Active against Gram-positive and Gram-negative bacteria, yeast, filamentous fungi, protozoa and insects. Potent bacterial and yeast RNA polymerases inhibitor. Inhibitor of fungal mannan and glucan formation. Similar to thiolutin. Antitumor compound.