Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aurintricarboxylic acid | 4431-00-9 | 100 MG
SDP

Catalog No. 5000230773
Encompass_Preferred

Aurintricarboxylic acid is a polyaromatic tricarboxylic acid used as a biochemical research reagent. It acts as an allosteric antagonist at P2X1 and P2X3 receptors and has reported anti-influenza and topoisomerase II inhibitory activity.

  • CAS 4431-00-9
  • Molecular formula: C22H14O9
  • Molecular weight: 422.34 g·mol⁻1
  • Appearance: solid powder
  • Storage: powder - -20°C (3 years) or 4°C (2 years)
  • Applications: receptor pharmacology and biochemical inhibition studies

Catalog No. 50-002-30773 Supplier Medchemexpress LLC Supplier No. HY122575100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aurintricarboxylic acid is a polyaromatic tricarboxylic acid used as a biochemical research reagent. It acts as an allosteric antagonist at P2X1 and P2X3 receptors and has reported anti-influenza and topoisomerase II inhibitory activity.

  • CAS 4431-00-9
  • Molecular formula: C22H14O9
  • Molecular weight: 422.34 g·mol⁻1
  • Appearance: solid powder
  • Storage: powder - -20°C (3 years) or 4°C (2 years)
  • Applications: receptor pharmacology and biochemical inhibition studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.