Learn More
Electron Microscopy Sciences Aurion Gold Sol, 10 nm 100 ML
SDP
Catalog No. 5026325
Encompass_Preferred
To prepare a high quality (immuno)gold conjugate it is important to have particles with uniform size and highly adsorptive surfaces. Prepared according to unique production protocols, Aurion Sols have the same narrow size distribution and adsorption properties as employed in the Conventional Immunogold Reagents.
The gold particles with a diameter suited for direct electron microscopic visualization. Aurion offers a range of Conventional Immunogold Reagents which cover the majority of approaches in transmission and scanning electron microscopy.
Aurion Gold Sols (solutions of high quality unconjugated gold particles) provide opportunities for users to prepare conjugates with primary antibodies, ligands and other binding agents with the same particle characteristics as in the Conventional Immunogold Reagents.
Particle size: 10 nm
Catalog No. 50-263-25 Supplier Electron Microscopy Sciences Supplier No. 25512
To prepare a high quality (immuno)gold conjugate it is important to have particles with uniform size and highly adsorptive surfaces. Prepared according to unique production protocols, Aurion Sols have the same narrow size distribution and adsorption properties as employed in the Conventional Immunogold Reagents.
The gold particles with a diameter suited for direct electron microscopic visualization. Aurion offers a range of Conventional Immunogold Reagents which cover the majority of approaches in transmission and scanning electron microscopy.
Aurion Gold Sols (solutions of high quality unconjugated gold particles) provide opportunities for users to prepare conjugates with primary antibodies, ligands and other binding agents with the same particle characteristics as in the Conventional Immunogold Reagents.
Particle size: 10 nm