Learn More
Electron Microscopy Sciences Aurion Aurion BSA-Fragment/Gold, UltraSmall 20 ML
SDP
Supplier:  Electron Microscopy Sciences 25498
Encompass_Preferred
Aurion's Bovine Serum Albumin is obtained from healthy livestock. BSA should be dissolved in an appropriate buffer, such as phosphate buffered saline, taking care not to denature the protein by foaming. The addition of BSA may cause a drop in pH of the final solution and correction may be required. As a preservative the use of NaN­3 or Kathon CG is recommended.
Particle size: UltraSmall
Catalog No. 50-245-82