Learn More
Electron Microscopy Sciences Aurion Mouse-Anti-FITC, EM-grade 10nm 1 ML
SDP
Catalog No. 5027428
Encompass_Preferred
For the electron microscopic visualization of FITC labels used in light microscopic specimens. For retrieval of faded FITC in DNA/RNA probes, fatty acids, and proteins.
Particle size: EM-grade 10nm
Catalog No. 50-274-28 Supplier Electron Microscopy Sciences Supplier No. 25583
For the electron microscopic visualization of FITC labels used in light microscopic specimens. For retrieval of faded FITC in DNA/RNA probes, fatty acids, and proteins.
Particle size: EM-grade 10nm