Learn More
Electron Microscopy Sciences Aurion Normal Donkey Serum 5 ML
SDP
Catalog No. 5027505
Encompass_Preferred
Normal sera are used to counteract the non-specific interaction between the sample and immunoglobulins. They can be added to the blocking solution and the incubation media. As a rule the normal serum species should be the same as the secondary antibody species (use normal goat serum with goat-anti-rabbit conjugates). Note: normal sera should not be used in combination with Protein A and Protein G gold reagents.
Catalog No. 50-275-05 Supplier Electron Microscopy Sciences Supplier No. 25572
Normal sera are used to counteract the non-specific interaction between the sample and immunoglobulins. They can be added to the blocking solution and the incubation media. As a rule the normal serum species should be the same as the secondary antibody species (use normal goat serum with goat-anti-rabbit conjugates). Note: normal sera should not be used in combination with Protein A and Protein G gold reagents.