Learn More
Electron Microscopy Sciences Aurion SE-EM Kit 90 ML
SDP
Catalog No. 5024585
Encompass_Preferred
AURION R-Gent SE-EM is a high efficiency silver enhancement reagent for electron microscopy. It intensifies the gold particles by homogeneous deposition of metallic silver on the particle surface. The resulting electron dense signal is easily detected and is compatible with heavy metal counterstaining. The reagent has extremely delayed auto-nucleation and can be used under standard laboratory light conditions. It also has low viscosity, which is especially advantageous for pre-embedding immunogold applications.
Catalog No. 50-245-85 Supplier Electron Microscopy Sciences Supplier No. 2552190
AURION R-Gent SE-EM is a high efficiency silver enhancement reagent for electron microscopy. It intensifies the gold particles by homogeneous deposition of metallic silver on the particle surface. The resulting electron dense signal is easily detected and is compatible with heavy metal counterstaining. The reagent has extremely delayed auto-nucleation and can be used under standard laboratory light conditions. It also has low viscosity, which is especially advantageous for pre-embedding immunogold applications.