Learn More
Medchemexpress LLC Auristatin E | 160800-57-7 | 99.9% | 732.01 | C40H69N5O7 | 100 MG
SDP
Catalog No. 5000204401
Encompass_Preferred
Auristatin E is a cytotoxic microtubule polymerization inhibitor used as a research reagent and as a payload in antibody-drug conjugate (ADC) studies. It blocks tubulin polymerization, inhibiting cell division and exhibiting potent antitumor activity in vitro.
  • Inhibits tubulin polymerization
  • Commonly used as an ADC payload in preclinical research
  • Provided as a solid reagent for in vitro applications
  • Contains CAS 160800-57-7 and molecular weight 732.01
Catalog No. 50-002-04401 Supplier Medchemexpress LLC Supplier No. HY15582100MG
Auristatin E is a cytotoxic microtubule polymerization inhibitor used as a research reagent and as a payload in antibody-drug conjugate (ADC) studies. It blocks tubulin polymerization, inhibiting cell division and exhibiting potent antitumor activity in vitro.
  • Inhibits tubulin polymerization
  • Commonly used as an ADC payload in preclinical research
  • Provided as a solid reagent for in vitro applications
  • Contains CAS 160800-57-7 and molecular weight 732.01