Learn More
Medchemexpress LLC Auristatin E | 160800-57-7 | MFCD25976744 | 99.9% | 732.01 g/mol | C40H69N5O7 | 250 MG
SDP
Catalog No. 5000204471
Encompass_Preferred
Auristatin E is a synthetic cytotoxic peptide that inhibits microtubule polymerization and is used as a payload in antibody-drug conjugate research. It is an analogue of dolastatin 10 and blocks cell division by preventing tubulin polymerization.
  • High purity suitable for research applications.
  • Inhibits tubulin polymerization to block cell division.
  • Used as a cytotoxic payload in antibody-drug conjugate research.
  • Contains CAS number 160800-57-7 and molecular weight 732.01 g/mol.
  • Available in laboratory pack sizes, including a 250 mg option.
Catalog No. 50-002-04471 Supplier Medchemexpress LLC Supplier No. HY15582250MG
Auristatin E is a synthetic cytotoxic peptide that inhibits microtubule polymerization and is used as a payload in antibody-drug conjugate research. It is an analogue of dolastatin 10 and blocks cell division by preventing tubulin polymerization.
  • High purity suitable for research applications.
  • Inhibits tubulin polymerization to block cell division.
  • Used as a cytotoxic payload in antibody-drug conjugate research.
  • Contains CAS number 160800-57-7 and molecular weight 732.01 g/mol.
  • Available in laboratory pack sizes, including a 250 mg option.