Learn More
Cayman Chemical Aurantioobtusin, Purity: ≥95%, CAS: 67979-25-3, 10 mg
SDP
Catalog No. 502941177
Encompass_Preferred
An anthraquinone with diverse biological activities; inhibits RLAR in vitro (IC50 = 13.6 μM); activates the AhR at 10 and 100 μM; inhibits arachidonic acid-, ADP-, and collagen-induced rat platelet aggregation; induces vasorelaxation in isolated precontracted rat mesenteric artery rings
Catalog No. 50-294-1177 Supplier Cayman Chemical Supplier No. 2748110 MG
An anthraquinone with diverse biological activities; inhibits RLAR in vitro (IC50 = 13.6 μM); activates the AhR at 10 and 100 μM; inhibits arachidonic acid-, ADP-, and collagen-induced rat platelet aggregation; induces vasorelaxation in isolated precontracted rat mesenteric artery rings