Learn More
AdipoGen Aurodox (2.5 mg)
SDP
Supplier:  AdipoGen AGCN20133MM25
Encompass_Preferred
Chemical. CAS: 12704-90-4. Formula: C44H62N2O12. MW: 811. Isolated from Streptomyces sp. K06-0806. Antibiotic Protein biosynthesis inhibitor by binding to elongation factor Tu (EF-Tu). Type 3 secretion system (T3SS) inhibitor. Growth promotor in poultry.
Catalog No. 50-205-3508