Learn More
Selleck Chemical LLC Aurora A Inhibitor I (TC-S 7010) 50mg 1158838-45-9
SDP
Catalog No. 507900
Encompass_Preferred
Aurora A Inhibitor I (TC-S 7010) is a novel, potent, and selective inhibitor of Aurora A with IC50 of 3.4 nM in a cell-free assay. It is 1000-fold more selective for Aurora A than Aurora B. Aurora A Inhibitor I (TC-S 7010) triggers apoptosis through the ROS-mediated UPR signaling pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-790-0 Supplier Selleck Chemical LLC Supplier No. S145150MG
Aurora A Inhibitor I (TC-S 7010) is a novel, potent, and selective inhibitor of Aurora A with IC50 of 3.4 nM in a cell-free assay. It is 1000-fold more selective for Aurora A than Aurora B. Aurora A Inhibitor I (TC-S 7010) triggers apoptosis through the ROS-mediated UPR signaling pathway. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.