Learn More
Cell Signaling Technology Aurora Antibody Sampler Kit, Unconjugated, 1 Kit
SDP
Catalog No. 502051187
Encompass
The Aurora Antibody Sampler Kit provides an economical means to investigate the G2/M phase of the cell cycle. The kit contains a selection of primary antibodies against Aurora kinase family members, including phospho-specific and total protein targets, along with a secondary antibody to facilitate western blot analysis.
  • Includes primary antibodies for Aurora A, Aurora B, and Aurora C, as well as specific phospho-targets at Thr198, Thr232, and Thr288.
  • Supplied with an HRP-linked anti-rabbit secondary antibody.
  • Validated for applications including western blotting, immunofluorescence, flow cytometry, and immunoprecipitation.
  • Storage conditions: –20°C in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Supports the investigation of mitotic structure, centrosome duplication, and chromosome segregation.
Catalog No. 50-205-1187 Supplier Cell Signaling Technology Supplier No. 3875T
The Aurora Antibody Sampler Kit provides an economical means to investigate the G2/M phase of the cell cycle. The kit contains a selection of primary antibodies against Aurora kinase family members, including phospho-specific and total protein targets, along with a secondary antibody to facilitate western blot analysis.
  • Includes primary antibodies for Aurora A, Aurora B, and Aurora C, as well as specific phospho-targets at Thr198, Thr232, and Thr288.
  • Supplied with an HRP-linked anti-rabbit secondary antibody.
  • Validated for applications including western blotting, immunofluorescence, flow cytometry, and immunoprecipitation.
  • Storage conditions: –20°C in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide.
  • Supports the investigation of mitotic structure, centrosome duplication, and chromosome segregation.