Learn More
Eurofins Discoverx Aurora B, active, 10 µg. (14-835)
Supplier:  Eurofins Discoverx 14835
Encompass
6xHis-tagged recombinant, human full length Aurora B and N-terminal GST-tagged recombinant, human INCENP amino acids 821-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9375