Learn More
Sino Biological Recombinant Human STRN & ALK Protein (GST Tag), Active
Catalog No. 502575922
Encompass_Preferred
Recombinant human fusion STRN (1-137a.a., exons1-3) - ALK (1058a.a.-end, exons20-29) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5922 Supplier Sino Biological Supplier No. A1919JG500
Recombinant human fusion STRN (1-137a.a., exons1-3) - ALK (1058a.a.-end, exons20-29) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.