Learn More
Cayman Chemical Aurora B KInaseNTRm Rabb 100uL
SDP
Supplier:  Cayman Chemical 32228100 UL
Encompass_Preferred
A DGK inhibitor (Ki 58 M) reduces basal and thrombin-stimulated phosphatidic acid levels in washed isolated human platelets at 30 100 and 500 M
Catalog No. 50-294-7737