Learn More
Enzo Life Sciences Aurora B (pThr232) polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPCC203F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Aurora B phosphorylated at Thr232. Source: Purified from rabbit serum. Species reactivity: Human. Creossreactivity: Weakly cross-reacts with mouse, rat, bovine and pig Aurora B. Applications: IHC (PS), WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7269