Learn More
Enzo Life Sciences Aurora B (pThr232) polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPCC203D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Aurora B phosphorylated at Thr232. UniProt ID: Q96GD4. Source: Purified from rabbit serum. Species reactivity: Human. Creossreactivity: Weakly cross-reacts with mouse, rat, bovine and pig Aurora B. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7268