Learn More
Aurora Microplates AURORA BLACK 384 ROUND WELL, 1
Catalog No. NC2151045
Encompass
Aurora Black 384 round well, 188uM bottom, non-barcoded, CO2 treated, black lid, std packaging, non-sterile 40/CS
This item is not returnable. View return policy
Catalog No. NC2151045 Supplier Aurora Microplates Supplier No. ABA201100A
Aurora Black 384 round well, 188uM bottom, non-barcoded, CO2 treated, black lid, std packaging, non-sterile 40/CS