Learn More
Medchemexpress LLC Aurora kinase inhibitor-2 | 331770-21-9 | 99.1% | 400.43 | 10 MG
SDP
Catalog No. 5000435691
Encompass_Preferred
Aurora kinase inhibitor-2 is a selective and ATP-competitive Aurora kinase inhibitor. It shows potent inhibition of Aurora A and Aurora B enzymes.
  • Selective and ATP-competitive Aurora kinase inhibitor.
  • IC50s of 310 nM for Aurora A and 240 nM for Aurora B.
  • Effective in an MCF7 cellular anti-proliferative assay with an IC50 of 1.25 μM.
Catalog No. 50-004-35691 Supplier Medchemexpress LLC Supplier No. HY11235510MG
Aurora kinase inhibitor-2 is a selective and ATP-competitive Aurora kinase inhibitor. It shows potent inhibition of Aurora A and Aurora B enzymes.
  • Selective and ATP-competitive Aurora kinase inhibitor.
  • IC50s of 310 nM for Aurora A and 240 nM for Aurora B.
  • Effective in an MCF7 cellular anti-proliferative assay with an IC50 of 1.25 μM.