Learn More
Medchemexpress LLC HY-112373 1mg Medchemexpress, Aurora Kinase Inhibitor 3 CAS:879127-16-9 Purity:>98%
SDP
Catalog No. 501969084
Encompass_Preferred
Medchemexpress, HY-112373 1mg Aurora Kinase Inhibitor 3 CAS:879127-16-9 Aurora Kinase Inhibitor 3 is a strong and selective Aurora A kinase inhibitor with an IC50 of 42 nM, and weakly inhibits EGFR with an IC50>10 μM. Aurora Kinase Inhibitor 3 has a binding mode with the cyclopropanecarboxylic acid moiety directed towards the solvent exposed region of the ATP-binding pocket, and several induced structural changes in the active-site compared with other published AIK structures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9084 Supplier Medchemexpress LLC Supplier No. HY1123731MG
Medchemexpress, HY-112373 1mg Aurora Kinase Inhibitor 3 CAS:879127-16-9 Aurora Kinase Inhibitor 3 is a strong and selective Aurora A kinase inhibitor with an IC50 of 42 nM, and weakly inhibits EGFR with an IC50>10 μM. Aurora Kinase Inhibitor 3 has a binding mode with the cyclopropanecarboxylic acid moiety directed towards the solvent exposed region of the ATP-binding pocket, and several induced structural changes in the active-site compared with other published AIK structures. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.