Learn More
Medchemexpress LLC Aurora kinase inhibitor II | 331770-21-9 | 99.1% | 400.43 g·mol⁻1 | C23H20N4O3 | 5 MG
SDP
Catalog No. 5000263653
Encompass_Preferred
Aurora kinase inhibitor-2 is a selective, ATP-competitive small-molecule inhibitor of Aurora kinases A and B used in biochemical and cell-based research. It shows inhibitory activity in the low hundreds of nanomolar range and is supplied as a solid suitable for in vitro and in vivo formulation studies.
  • Selective ATP-competitive inhibitor of Aurora A and Aurora B (IC50: Aurora A 310 nM; Aurora B 240 nM).
  • Supplied as a white to yellow solid with high reported purity (~99.1%).
  • Molecular formula C23H20N4O3 and molecular weight 400.43 g·mol⁻1.
  • Dissolves readily in DMSO; recommended in vitro solubility 100 mg/mL (ultrasonic may be required).
  • Formulatable for in vivo studies using common vehicles (SBE-β-CD, corn oil, PEG300/Tween-80 mixes).
  • Intended for research applications probing Aurora kinase function and signaling.
Catalog No. 50-002-63653 Supplier Medchemexpress LLC Supplier No. HY1123555MG
Aurora kinase inhibitor-2 is a selective, ATP-competitive small-molecule inhibitor of Aurora kinases A and B used in biochemical and cell-based research. It shows inhibitory activity in the low hundreds of nanomolar range and is supplied as a solid suitable for in vitro and in vivo formulation studies.
  • Selective ATP-competitive inhibitor of Aurora A and Aurora B (IC50: Aurora A 310 nM; Aurora B 240 nM).
  • Supplied as a white to yellow solid with high reported purity (~99.1%).
  • Molecular formula C23H20N4O3 and molecular weight 400.43 g·mol⁻1.
  • Dissolves readily in DMSO; recommended in vitro solubility 100 mg/mL (ultrasonic may be required).
  • Formulatable for in vivo studies using common vehicles (SBE-β-CD, corn oil, PEG300/Tween-80 mixes).
  • Intended for research applications probing Aurora kinase function and signaling.