Learn More
Apexbio Technology LLC Aurora Kinase Inhibitor III 879127-16-9 5mg
SDP
Catalog No. 5000088140
Encompass_Preferred
Aurora Kinase Inhibitor III (CAS 879127-16-9) is a selective small molecule inhibitor targeting Aurora A kinase It acts by binding to the ATP-binding site of Aurora A thereby suppressing its enzymatic activity with an IC50 of 42 nM The compound exhibits substantially lower inhibitory potency against epidermal growth factor receptor (EGFR) with an IC50 greater than 10 M Aurora Kinase Inhibitor III serves as a useful chemical probe for investigating Aurora A-mediated processes including mitotic regulation cell cycle progression and associated signaling pathways and supports studies exploring the role of Aurora A in oncogenesis
Catalog No. 50-000-88140 Supplier Apexbio Technology LLC Supplier No. B79475
Aurora Kinase Inhibitor III (CAS 879127-16-9) is a selective small molecule inhibitor targeting Aurora A kinase It acts by binding to the ATP-binding site of Aurora A thereby suppressing its enzymatic activity with an IC50 of 42 nM The compound exhibits substantially lower inhibitory potency against epidermal growth factor receptor (EGFR) with an IC50 greater than 10 M Aurora Kinase Inhibitor III serves as a useful chemical probe for investigating Aurora A-mediated processes including mitotic regulation cell cycle progression and associated signaling pathways and supports studies exploring the role of Aurora A in oncogenesis