Learn More
Cayman Chemical Aurora Kinase Inhibitor III, Purity: ≥98%, CAS: 879127-16-9, 5 mg
SDP
Catalog No. 502986124
Encompass_Preferred
A potent inhibitor of Aurora A kinase (IC50 = 42 nM); selective for Aurora A over BMX, BTK, IGF-1R, c-Src, TRKB, SYK, and EGFR (IC50s = 386, 3,550, 591, 1,980, 2,510, 887, and >10,000 nM, respectively)
Catalog No. 50-298-6124 Supplier Cayman Chemical Supplier No. 218515 MG
A potent inhibitor of Aurora A kinase (IC50 = 42 nM); selective for Aurora A over BMX, BTK, IGF-1R, c-Src, TRKB, SYK, and EGFR (IC50s = 386, 3,550, 591, 1,980, 2,510, 887, and >10,000 nM, respectively)