Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 9212MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: WB. Conjugate/Tag: Unconjugated. Clone: AURKB/1521. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: HeLa cells. Tonsil, Placenta, Hodgkin s Lymphoma or Colon Carcinoma. Cellular Staining: Nuclear and Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human Aurora B protein fragment (aa89-251). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 39kDa.Chromosome: 17p13.1. Gene Symbol: AURKB. Gene ID: 9212. Ensemble: ENSG00000178999. Uniprot: NP_001300884.
Catalog No. 50-188-5323