Learn More
Medchemexpress LLC Aurothiomalate tetramer (sodium) | 12244-57-4 | MFCD00064304 | 98.0% | 390.08 g·mol-1 | C4H6O4S·Au·xNa | 25 MG
SDP
Catalog No. 5000242534
Encompass_Preferred
Aurothiomalate sodium is a gold-containing anti-rheumatic reagent used in research to inhibit PKCι signaling and thioredoxin reductase. Supplied as a powder for biochemical and cell-based studies, it is used to probe redox biology and oncogenic signaling pathways.
  • Purity: 98.0%
  • Gold-containing thiomalate compound for biochemical research
  • Inhibits PKCι signaling and thioredoxin reductase
  • Suitable for cell-based assays and mechanistic studies
  • Available in small milligram pack sizes for laboratory use
  • Store according to the safety data sheet to preserve stability
Catalog No. 50-002-42534 Supplier Medchemexpress LLC Supplier No. HY10638125MG
Aurothiomalate sodium is a gold-containing anti-rheumatic reagent used in research to inhibit PKCι signaling and thioredoxin reductase. Supplied as a powder for biochemical and cell-based studies, it is used to probe redox biology and oncogenic signaling pathways.
  • Purity: 98.0%
  • Gold-containing thiomalate compound for biochemical research
  • Inhibits PKCι signaling and thioredoxin reductase
  • Suitable for cell-based assays and mechanistic studies
  • Available in small milligram pack sizes for laboratory use
  • Store according to the safety data sheet to preserve stability