Learn More
Bio Rad Laboratories AURUM AEX MINI KIT
Catalog No. 509011889
Encompass
Mini kit, Anion exchange (AEX); Bio-Rad; Aurum; UNOsphere Q; support; Binding capacity: 36 mg protein/column; Column bed 0.2mL; Binding buffer: 20mm Tris, pH 8.3; Elution buffer: binding buffer + 1.0M NaCl; 25/Pk
Catalog No. 50-901-1889 Supplier Bio Rad Laboratories Supplier No. 7326706
Mini kit, Anion exchange (AEX); Bio-Rad; Aurum; UNOsphere Q; support; Binding capacity: 36 mg protein/column; Column bed 0.2mL; Binding buffer: 20mm Tris, pH 8.3; Elution buffer: binding buffer + 1.0M NaCl; 25/Pk