Learn More
Alkali Scientific Autoclave Sterilization Bags, Red, 12.2 x 26" (31 x 66cm) 200/cs
SDP
Catalog No. 502336131
Encompass_Preferred
The autoclave sterilization bags are one of the best products used to isolate and store hazardous waste in various research laboratories. The heavy gauge polyethylene material offers the best protection due to its superior puncture-resistance. This material allows the autoclave sterilization bags to contain waste at temperatures of up to 136 Degrees Celsius. Autoclave sterilization bags have a white marking panel that allows quick and easy to spot labelling. Labelling can be done with any type of marker or permanent marker. The white marking panel is very noticeable due to its color that offers background contrast and also due its size and location. The words “Hazardous Waste” and the “Biohazard” symbol appears on each one of the autoclave sterilization bags in a visible way. This ensures that accidents are reduced to a minimum and conforms with the international standards for biohazard manipulation - OSHA standard 29CFR191. 200 per pack.
Catalog No. 50-233-6131 Supplier Alkali Scientific Supplier No. A3000R
The autoclave sterilization bags are one of the best products used to isolate and store hazardous waste in various research laboratories. The heavy gauge polyethylene material offers the best protection due to its superior puncture-resistance. This material allows the autoclave sterilization bags to contain waste at temperatures of up to 136 Degrees Celsius. Autoclave sterilization bags have a white marking panel that allows quick and easy to spot labelling. Labelling can be done with any type of marker or permanent marker. The white marking panel is very noticeable due to its color that offers background contrast and also due its size and location. The words “Hazardous Waste” and the “Biohazard” symbol appears on each one of the autoclave sterilization bags in a visible way. This ensures that accidents are reduced to a minimum and conforms with the international standards for biohazard manipulation - OSHA standard 29CFR191. 200 per pack.