Learn More
Research Products International Corp Autoclave Bags, 25""x35"", 1.75 mil, 200/pk
SDP
Catalog No. 502229766
Encompass
Heat resistant bag is designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Clear bags allow full visibility of medical supplies and supplies. Supplied 200 per package.
Catalog No. 50-222-9766 Supplier Research Products International Corp Supplier No. 248257
Heat resistant bag is designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Clear bags allow full visibility of medical supplies and supplies. Supplied 200 per package.