Learn More
Research Products International Corp Autoclave Bags, 30""x34"", 2mil, 200/pk
SDP
Catalog No. 502229767
Encompass
Heat resistant bags designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Clear bags allow full visibility of medical supplies and supplies. Supplied 100 per package.
Catalog No. 50-222-9767 Supplier Research Products International Corp Supplier No. 248255CTN
Heat resistant bags designed for all autoclaving procedures. High density bags are stronger than regular polypropylene bags for sterilizing equipment and supplies. Clear bags allow full visibility of medical supplies and supplies. Supplied 100 per package.