Learn More
New England Biolabs, Inc. Authenticase - 125 reactions
SDP
Supplier:  New England Biolabs, Inc. M0689L
Encompass
Authenticase is a mixture of enzymes capable of recognizing mismatch and indel regions on a double-stranded DNA and creating double-stranded cleavages around these regions and with a limited non-specific activity on homoduplex regions of DNA. The regions of mismatch or indel could range from at least 1 to 10 bps. The activity of cleavage on mismatch/Indel regions can be utilized in the process of oligo-based PCR gene synthesis to improve the accuracy of PCR amplicons. Alternatively, Authenticase treated products can be analyzed to determine the popularity of heteroduplex of a PCR pool amplified from enzyme-based edited genome.
Catalog No. 50-252-7888