Learn More
Proteintech Group Inc AuthentiKine™ Human IL-6 ELISA Kit
SDP
Supplier:  Proteintech Group Inc KE0013996T
Encompass_Preferred
KE00139 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The IL6 ELISA kit is to be used to detect and quantify protein levels of endogenous IL6. The assay recognizes human IL6. An antibody specific for IL6 has been pre-coated onto the microwells. The IL6 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for IL6 is added to detect the captured IL6 protein. For signal development, Sterptravidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Catalog No. 50-225-8397