Learn More
Proteintech Group Inc AuthentiKine™ Human TNF-alpha ELISA Kit
SDP
Supplier:  Proteintech Group Inc KE0015496T
Encompass_Preferred
KE00154 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The TNF-alpha ELISA kit is to be used to detect and quantify protein levels of endogenous TNF-alpha. The assay recognizes human TNF-alpha. An antibody specific for TNF-alpha has been pre-coated onto the microwells. The TNF-alpha protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody specific for TNF-alpha is added to detect the captured TNF-alpha protein. For signal development, horseradish peroxidase (HRP)-conjugated antibody is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.
Catalog No. 50-225-8400