Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray AUTION Check Plus Control Solution
SDP

Catalog No. 21403495
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-403-495 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning: For in vitro diagnostic use only. Handle according to laboratory safety guidelines. Avoid contact with skin or eyes. Store as directed and use protective equipment when handling.
Catalog No. 21-403-495 Supplier Arkray Supplier No. 102478
Add to Cart
Add to Cart
Warning: For in vitro diagnostic use only. Handle according to laboratory safety guidelines. Avoid contact with skin or eyes. Store as directed and use protective equipment when handling.

This Control Solution provides accurate and reliable quality control for A1c analyzers, ensuring consistent results, easy workflow integration, and confidence in every lab test.

  • Multi-Level Control: Provides low, normal, and high levels to verify analyzer accuracy across clinically relevant ranges.
  • Easy Integration: Seamlessly fits into daily laboratory workflows for fast and convenient quality checks.
  • Stable and Consistent

Specifications

Sample Type Urine
Type Control Solution
Level 1, 2
Quantity 25 mL
Form Liquid

AUTION CHECK PLUS Control Solution provides consistent, accurate quality control for A1c analyzers, supporting reliable lab results and efficient workflow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.